Learn More
Selleck Chemical LLC L-Ornithine hydrochloride S4653-100mg
SDP
Catalog No. 503111379
Encompass_Preferred
L-ornithine hydrochloride ((S)-2 5-Diaminopentanoic acid) has an antifatigue effect by increasing the efficiency of energy consumption and promoting the excretion of ammonia It is one of the key reactants in the urea cycle
Catalog No. 50-311-1379 Supplier Selleck Chemical LLC Supplier No. S4653100MG
L-ornithine hydrochloride ((S)-2 5-Diaminopentanoic acid) has an antifatigue effect by increasing the efficiency of energy consumption and promoting the excretion of ammonia It is one of the key reactants in the urea cycle