Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine (hydrochloride) | 3184-13-2 | MFCD00064562 | 99.6% | 168.62 g·mol⁻¹ | C5H13ClN2O2 | 1 ML
SDP

Catalog No. 5000301032
Encompass_Preferred

L-ornithine hydrochloride is a non-proteinogenic amino acid used in urea cycle and nitrogen metabolism research. Intended for laboratory and analytical applications, the material is supplied as a solid powder and as a ready-to-use aqueous solution; supporting documents such as SDS and COA are provided by the manufacturer.

  • Non-proteinogenic amino acid used in urea cycle and nitrogen metabolism studies.
  • Available as solid powder and as a 10 mM aqueous solution (1 mL).
  • High purity (99.6% assay by qNMR).
  • Molecular weight 168.62 g·mol⁻¹; formula C5H13ClN2O2.
  • SDS and certificate of analysis available for safety and quality information.

Catalog No. 50-003-01032 Supplier Medchemexpress LLC Supplier No. HYW01701810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ornithine hydrochloride is a non-proteinogenic amino acid used in urea cycle and nitrogen metabolism research. Intended for laboratory and analytical applications, the material is supplied as a solid powder and as a ready-to-use aqueous solution; supporting documents such as SDS and COA are provided by the manufacturer.

  • Non-proteinogenic amino acid used in urea cycle and nitrogen metabolism studies.
  • Available as solid powder and as a 10 mM aqueous solution (1 mL).
  • High purity (99.6% assay by qNMR).
  • Molecular weight 168.62 g·mol⁻¹; formula C5H13ClN2O2.
  • SDS and certificate of analysis available for safety and quality information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.