Learn More
Medchemexpress LLC L-ornithine hydrochloride | 3184-13-2 | MFCD00064562 | 99.6% | 168.62 g/mol | C5H13ClN2O2 | 5 G
SDP
Catalog No. 5000301266
Encompass_Preferred
L-ornithine hydrochloride is a non-proteinogenic amino acid used in research to study the urea cycle and nitrogen metabolism. Supplied as a white to off-white solid, it is commonly used in biochemical assays, analytical standards, and cell-culture applications where high-purity ornithine is required.
  • Supports studies of the urea cycle and nitrogen disposal.
  • High-purity material suitable for analytical and research use.
  • Available as a solid for easy weighing and solution preparation.
  • Provided with a certificate of analysis for batch-specific characterization.
  • Applicable to metabolic and nephroprotection research.
Catalog No. 50-003-01266 Supplier Medchemexpress LLC Supplier No. HYW0170185G
L-ornithine hydrochloride is a non-proteinogenic amino acid used in research to study the urea cycle and nitrogen metabolism. Supplied as a white to off-white solid, it is commonly used in biochemical assays, analytical standards, and cell-culture applications where high-purity ornithine is required.
  • Supports studies of the urea cycle and nitrogen disposal.
  • High-purity material suitable for analytical and research use.
  • Available as a solid for easy weighing and solution preparation.
  • Provided with a certificate of analysis for batch-specific characterization.
  • Applicable to metabolic and nephroprotection research.