Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine (hydrochloride) | 3184-13-2 | MFCD00064562 | 99.9% | 168.62 | C5H13ClN2O2 | 50 MG
SDP

Catalog No. 5000301267
Encompass_Preferred

L-Ornithine hydrochloride is the hydrochloride salt of the non-proteinogenic amino acid L-ornithine. It is used as a biochemical reagent and analytical standard in research on the urea cycle, nitrogen metabolism, and amino acid analysis.

  • High purity (99.85%), suitable for analytical use.
  • Hydrochloride salt form for enhanced stability and solubility in aqueous solutions.
  • Commonly used in urea cycle and nitrogen metabolism studies.
  • Available in small research quantities for laboratory experiments.
  • Molecular weight 168.62; formula C5H13ClN2O2.

Catalog No. 50-003-01267 Supplier Medchemexpress LLC Supplier No. HYW017018R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ornithine hydrochloride is the hydrochloride salt of the non-proteinogenic amino acid L-ornithine. It is used as a biochemical reagent and analytical standard in research on the urea cycle, nitrogen metabolism, and amino acid analysis.

  • High purity (99.85%), suitable for analytical use.
  • Hydrochloride salt form for enhanced stability and solubility in aqueous solutions.
  • Commonly used in urea cycle and nitrogen metabolism studies.
  • Available in small research quantities for laboratory experiments.
  • Molecular weight 168.62; formula C5H13ClN2O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.