Learn More
Medchemexpress LLC L-ornithine hydrochloride | 3184-13-2 | MFCD00064562 | 99.9% | 168.62 g·mol⁻1 | C5H13ClN2O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW017018500MG
Encompass_Preferred
L-ornithine hydrochloride is a research-grade, non-proteinogenic amino acid used in studies of nitrogen metabolism and the urea cycle. Supplied as a white to off-white solid, the material is provided with batch-specific quality documentation for laboratory and analytical use.
  • High purity (~99.9%) for research and analytical applications.
  • Used in urea cycle and nitrogen metabolism studies.
  • White to off-white solid, facile handling and weighing.
  • Available in small laboratory sizes, including 500 mg.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-003-01033