Learn More
Medchemexpress LLC L-ornithine (hydrochloride) | 3184-13-2 | MFCD00064562 | 99.9% | 168.62 g/mol | C5H13ClN2O2 | 100 MG
SDP
Catalog No. 5000300655
Encompass_Preferred
L-Ornithine hydrochloride (CAS 3184-13-2) is an analytical standard of the non-proteinogenic amino acid L-ornithine. Supplied as a white to off-white solid with high assay value, it is intended for method development, calibration, and qualitative and quantitative analysis in chromatographic and mass spectrometric workflows.
  • High purity (99.85%) for accurate quantitation.
  • Suitable for HPLC, GC, and MS applications.
  • White to off-white solid form for easy handling.
  • Used as a reference standard in analytical methods.
  • Room temperature shipping and standard storage recommendations apply.
  • Provided with safety data sheet and certificate of analysis.
Catalog No. 50-003-00655 Supplier Medchemexpress LLC Supplier No. HYW017018B100MG
L-Ornithine hydrochloride (CAS 3184-13-2) is an analytical standard of the non-proteinogenic amino acid L-ornithine. Supplied as a white to off-white solid with high assay value, it is intended for method development, calibration, and qualitative and quantitative analysis in chromatographic and mass spectrometric workflows.
  • High purity (99.85%) for accurate quantitation.
  • Suitable for HPLC, GC, and MS applications.
  • White to off-white solid form for easy handling.
  • Used as a reference standard in analytical methods.
  • Room temperature shipping and standard storage recommendations apply.
  • Provided with safety data sheet and certificate of analysis.