Learn More
Medchemexpress LLC L-ornithine (hydrochloride) | 3184-13-2 | MFCD00064562 | 99.9% | 168.62 g/mol | C5H13ClN2O2 | 25 MG
SDP
Catalog No. 5000303033
Encompass_Preferred
L-ornithine (hydrochloride) is the monohydrochloride salt of the amino acid L-ornithine, provided as a research-grade analytical standard for biochemical and analytical applications. It is commonly used in studies of the urea cycle and amino acid metabolism, and as a reference material for analytical assays.
  • High purity suitable for analytical use.
  • Available in small, measured vial sizes for standards and assays.
  • Consistent lot-to-lot quality for reproducible results.
  • Appropriate for biochemical and metabolic studies involving the urea cycle.
  • Supplied with safety data sheet and product specifications.
Catalog No. 50-003-03033 Supplier Medchemexpress LLC Supplier No. 25MGHYW017018R
L-ornithine (hydrochloride) is the monohydrochloride salt of the amino acid L-ornithine, provided as a research-grade analytical standard for biochemical and analytical applications. It is commonly used in studies of the urea cycle and amino acid metabolism, and as a reference material for analytical assays.
  • High purity suitable for analytical use.
  • Available in small, measured vial sizes for standards and assays.
  • Consistent lot-to-lot quality for reproducible results.
  • Appropriate for biochemical and metabolic studies involving the urea cycle.
  • Supplied with safety data sheet and product specifications.