Learn More
Medchemexpress LLC L-ornithine hydrochloride | 3184-13-2 | MFCD00064562, MFCD00166412 | 99.9% | 168.62 g/mol | C5H13ClN2O2 | 1 G
SDP
Catalog No. 5000299883
Encompass_Preferred
L-ornithine hydrochloride is a non-proteinogenic amino acid supplied for research use. It participates in the urea cycle to remove excess nitrogen in vivo and has reported nephroprotective activity, making it useful in biochemical and pharmacological studies.
  • Used in urea cycle and nitrogen metabolism research.
  • Reported nephroprotective properties in preclinical studies.
  • High purity (99.91%) suitable for biochemical assays.
  • Molecular formula C5H13ClN2O2; molecular weight 168.62 g/mol.
  • Available as a 1 g research pack.
Catalog No. 50-002-99883 Supplier Medchemexpress LLC Supplier No. HYW0170181G
L-ornithine hydrochloride is a non-proteinogenic amino acid supplied for research use. It participates in the urea cycle to remove excess nitrogen in vivo and has reported nephroprotective activity, making it useful in biochemical and pharmacological studies.
  • Used in urea cycle and nitrogen metabolism research.
  • Reported nephroprotective properties in preclinical studies.
  • High purity (99.91%) suitable for biochemical assays.
  • Molecular formula C5H13ClN2O2; molecular weight 168.62 g/mol.
  • Available as a 1 g research pack.