Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ornithine Hydrochloride, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611372 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-372 1 kg Poly Bottle
18-611-374 25 g Poly Bottle
18-611-373 100 g Poly Bottle
18-611-377 500 g Poly Bottle
18-611-376 5 kg Poly Bottle
18-611-375 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-372 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. OR1301KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 1 kg
Specific Rotation +23° to +25.0°
Percent Purity 0.99
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material L-Ornithine Hydrochloride

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.