Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca L-Ornithine Hydrochloride (Dietary Supplement Grade)
SDP

Catalog No. 14185528 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-528 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-528 Supplier Medisca Supplier No. 137009
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ornithine Hydrochloride (Dietary Supplement Grade) is a white crystalline powder that is soluble in water. If using for sterile preparation, ensure further processing and testing.

L-Ornithine Hydrochloride is a white crystalline powder that is soluble in water.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): L-Ornithine, Ornithine L-monohydrochloride

Specifications

Chemical Name or Material L-Ornithine Hydrochloride (Dietary Supplement Grade)
Packaging Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck
CAS 3184-13-2
Type Drug Standard
Molecular Weight (g/mol) 168.621
Quantity 1 kg
Percent Purity 99 to 101 %
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.