Learn More
Apexbio Technology LLC L-ORNITHINE-L-ASPARTATE-200MG
SDP
Catalog No. 5000570169
Encompass_Preferred
L-Ornithine L-aspartate (CAS No 3230-94-2) is a compound commonly used in biomedical research within the field of hepatology due to its potential in mitigating hepatic encephalopathy Originating from amino acids its mechanism of action involves enhancing ammonia detoxification through the urea cycle and glutamine synthesis in the liver It influences pathways related to nitrogen metabolism and liver function In vitro activity generally falls within the nanomolar to low micromolar range revealing its potent biological efficacy L-Ornithine L-aspartate is frequently studied in cell models and animal studies especially for its role in reducing ammonia levels and supporting liver function It is also utilized in drug screening processes for liver-related conditions Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70169 Supplier Apexbio Technology LLC Supplier No. C6859200
L-Ornithine L-aspartate (CAS No 3230-94-2) is a compound commonly used in biomedical research within the field of hepatology due to its potential in mitigating hepatic encephalopathy Originating from amino acids its mechanism of action involves enhancing ammonia detoxification through the urea cycle and glutamine synthesis in the liver It influences pathways related to nitrogen metabolism and liver function In vitro activity generally falls within the nanomolar to low micromolar range revealing its potent biological efficacy L-Ornithine L-aspartate is frequently studied in cell models and animal studies especially for its role in reducing ammonia levels and supporting liver function It is also utilized in drug screening processes for liver-related conditions Experimental concentrations typically depend on the specific research design