Learn More
Medchemexpress LLC L-Ornithine L-aspartate | 3230-94-2 | 98.0% | C9H19N3O6 | 1 G
SDP
Catalog No. 5000433450
Encompass_Preferred
L-Ornithine L-aspartate is a stable salt of two natural nonessential L-amino acids, ornithine and aspartic acid, with oral activity. It lowers blood ammonia concentration and helps eliminate symptoms of hepatic encephalopathy associated with liver cirrhosis, while also improving mitochondrial functions.
  • Lowers blood ammonia concentration.
  • Eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis.
  • Improves mitochondrial functions.
  • Increases redox activity of mitochondria in SH-SY5Y cells.
  • Counteracts ROT- and 6-OHDA-induced mitochondrial depolarization (In Vitro).
  • Prevents ammonia-precipitated coma in portal-systemic encephalopathy (PSE) rats (In Vivo).
  • Protects against Thioacetamide-induced hepatic damage in rats (In Vivo).
Catalog No. 50-004-33450 Supplier Medchemexpress LLC Supplier No. HYA02821G
L-Ornithine L-aspartate is a stable salt of two natural nonessential L-amino acids, ornithine and aspartic acid, with oral activity. It lowers blood ammonia concentration and helps eliminate symptoms of hepatic encephalopathy associated with liver cirrhosis, while also improving mitochondrial functions.
  • Lowers blood ammonia concentration.
  • Eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis.
  • Improves mitochondrial functions.
  • Increases redox activity of mitochondria in SH-SY5Y cells.
  • Counteracts ROT- and 6-OHDA-induced mitochondrial depolarization (In Vitro).
  • Prevents ammonia-precipitated coma in portal-systemic encephalopathy (PSE) rats (In Vivo).
  • Protects against Thioacetamide-induced hepatic damage in rats (In Vivo).