Learn More
Medchemexpress LLC L-Ornithine L-aspartate | 3230-94-2 | 98.0% | C9H19N3O6 | 1 ML
SDP
Catalog No. 5000432560
Encompass_Preferred
The product is a solution of L-Ornithine L-aspartate. It is a stable salt of two natural nonessential L-amino acids: ornithine and aspartic acid. This product is for research use only and is not sold to patients. It lowers blood ammonia concentration, eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis, and improves mitochondrial functions.
  • Increases mitochondrial redox activity in SH-SY5Y cells
  • Counteracts ROT- and 6-OHDA-induced mitochondrial depolarization
  • Prevents ammonia-precipitated coma in portal-systemic encephalopathy (PSE) rats
  • Protects against Thioacetamide-induced hepatic damage in rats
  • Used in research for inflammation or immune system disease
  • Used in research for metabolic or endocrine disease
  • Used in research for metabolic disease
Catalog No. 50-004-32560 Supplier Medchemexpress LLC Supplier No. HYA028210MM/1ML
The product is a solution of L-Ornithine L-aspartate. It is a stable salt of two natural nonessential L-amino acids: ornithine and aspartic acid. This product is for research use only and is not sold to patients. It lowers blood ammonia concentration, eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis, and improves mitochondrial functions.
  • Increases mitochondrial redox activity in SH-SY5Y cells
  • Counteracts ROT- and 6-OHDA-induced mitochondrial depolarization
  • Prevents ammonia-precipitated coma in portal-systemic encephalopathy (PSE) rats
  • Protects against Thioacetamide-induced hepatic damage in rats
  • Used in research for inflammation or immune system disease
  • Used in research for metabolic or endocrine disease
  • Used in research for metabolic disease