Learn More
Medchemexpress LLC L-Ornithine L-aspartate | 3230-94-2 | 98.0% | C9H19N3O6 | 5 G
SDP
Catalog No. 5000433341
Encompass_Preferred
L-Ornithine L-aspartate is a stable salt of two natural nonessential L-amino acids: ornithine and aspartic acid. It has oral activity and is known to lower blood ammonia concentration, help eliminate symptoms of hepatic encephalopathy associated with liver cirrhosis, and improve mitochondrial functions.
  • Lowers blood ammonia concentration
  • Eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis
  • Improves mitochondrial functions
  • Increases redox activity of mitochondria in SH-SY5Y cells
  • Prevents ammonia-precipitated coma in PSE rats
  • Protects against Thioacetamide-induced hepatic damage in rats
Catalog No. 50-004-33341 Supplier Medchemexpress LLC Supplier No. HYA02825G
L-Ornithine L-aspartate is a stable salt of two natural nonessential L-amino acids: ornithine and aspartic acid. It has oral activity and is known to lower blood ammonia concentration, help eliminate symptoms of hepatic encephalopathy associated with liver cirrhosis, and improve mitochondrial functions.
  • Lowers blood ammonia concentration
  • Eliminates symptoms of hepatic encephalopathy associated with liver cirrhosis
  • Improves mitochondrial functions
  • Increases redox activity of mitochondria in SH-SY5Y cells
  • Prevents ammonia-precipitated coma in PSE rats
  • Protects against Thioacetamide-induced hepatic damage in rats