Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ornithine L-Aspartate 96.0+%, TCI America™
SDP

Catalog No. O044025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O044025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O044025G Supplier TCI America Supplier No. O044025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3230-94-2
Molecular Formula C9H19N3O6
Molecular Weight (g/mol) 265.27
MDL Number MFCD00058084
InChI Key IXUZXIMQZIMPSQ-ZBRNBAAYSA-N
PubChem CID 10220941
IUPAC Name (4S)-4-amino-4-carboxybutan-1-aminium (2S)-2-amino-3-carboxypropanoate
SMILES N[C@@H](CCC[NH3+])C(O)=O.N[C@@H](CC(O)=O)C([O-])=O

Specifications

Melting Point 204°C
Color White
Quantity 25 g
Formula Weight 265.27
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Ornithine L-Aspartate
RTECSNumber CI9463000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.