Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ornithine methyl ester dihydrochloride, 97%

Catalog No. AAH6322522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6322522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6322522 Supplier Thermo Scientific Chemicals Supplier No. H6322522

missing translation for 'validMainframeMsg'

CAS: 40216-82-8 | C6H16Cl2N2O2 | 219.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40216-82-8
Molecular Formula C6H16Cl2N2O2
Molecular Weight (g/mol) 219.106
MDL Number MFCD00190977
InChI Key SNFZNVWAQDVFAK-XRIGFGBMSA-N
PubChem CID 14273060
IUPAC Name methyl (2S)-2,5-diaminopentanoate;dihydrochloride
SMILES COC(=O)C(CCCN)N.Cl.Cl

Specifications

Color White
Synonym h-orn-ome.2hcl, l-ornithine methyl ester dihydrochloride, methyl l-ornithine dihydrochloride, s-methyl 2,5-diaminopentanoate dihydrochloride, h-orn-ome?hcl, methyl l-ornithine hcl, h-orn-ome 2hcl, h-orn-ome2hcl, l-ornithine, methyl ester, dihydrochloride, h-orn-ome 2 hcl
Quantity 100 g
Formula Weight 219.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L-Ornithine methyl ester dihydrochloride
EINECSNumber 254-841-5
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.