Learn More
Medchemexpress LLC L-Ornithine monohydrochloride | 3184-13-2 | MFCD00012917 | 99.7% | 168.62 g/mol | C5H13ClN2O2 | 250 MG
SDP
Catalog No. 5000302352
Encompass_Preferred
L-Ornithine monohydrochloride is the hydrochloride salt of L-ornithine supplied as a high-purity, crystalline powder for laboratory and cell-culture use. It is used in studies of the urea cycle, amino-acid metabolism, and as a supplement in cell-culture media, and is suitable for analytical and biological applications.
  • High purity suitable for cell-culture applications.
  • Supplied as a stable, crystalline powder for easy handling and storage.
  • Supports urea cycle and amino-acid metabolism studies.
  • Compatible with common analytical techniques and buffer systems.
  • Available in small bench-scale pack sizes for research workflows.
Catalog No. 50-003-02352 Supplier Medchemexpress LLC Supplier No. 250MGHYW017018B
L-Ornithine monohydrochloride is the hydrochloride salt of L-ornithine supplied as a high-purity, crystalline powder for laboratory and cell-culture use. It is used in studies of the urea cycle, amino-acid metabolism, and as a supplement in cell-culture media, and is suitable for analytical and biological applications.
  • High purity suitable for cell-culture applications.
  • Supplied as a stable, crystalline powder for easy handling and storage.
  • Supports urea cycle and amino-acid metabolism studies.
  • Compatible with common analytical techniques and buffer systems.
  • Available in small bench-scale pack sizes for research workflows.