Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine monohydrochloride | 3184-13-2 | MFCD00064562 | 99.7% | 168.62 g/mol | C5H13ClN2O2 | 50 MG
SDP

Catalog No. 5000302052
Encompass_Preferred

L-ornithine monohydrochloride is a research-grade, high-purity amino acid salt commonly used in biochemical studies and cell culture. It participates in the urea cycle and serves as a precursor for arginine and polyamine biosynthesis. Supplied in small quantities for laboratory use, this reagent is intended for analytical, cell culture, and biochemical applications that require verified identity and purity.

  • High purity: 99.73%.
  • Molecular weight: 168.62 g/mol.
  • Chemical formula: C5H13ClN2O2.
  • CAS number: 3184-13-2.
  • Suitable for cell culture applications.
  • Available in small research sizes, including 50 mg.

Catalog No. 50-003-02052 Supplier Medchemexpress LLC Supplier No. HYW017018B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ornithine monohydrochloride is a research-grade, high-purity amino acid salt commonly used in biochemical studies and cell culture. It participates in the urea cycle and serves as a precursor for arginine and polyamine biosynthesis. Supplied in small quantities for laboratory use, this reagent is intended for analytical, cell culture, and biochemical applications that require verified identity and purity.

  • High purity: 99.73%.
  • Molecular weight: 168.62 g/mol.
  • Chemical formula: C5H13ClN2O2.
  • CAS number: 3184-13-2.
  • Suitable for cell culture applications.
  • Available in small research sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.