Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ornithine monohydrochloride, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111013914 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3914 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3914 Supplier MilliporeSigma Supelco Supplier No. 57197100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 3184-13-2
Linear Formula C5H12N2O2 · HCl
MDL Number MFCD00064562
Synonym (S)-2,5-Diaminopentanoic acid monohydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 168.62
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.