Learn More
Bellco Glass, Inc. Low Profile 2-P Stainless Steel Sparger Assembly , 6L
SDP
Catalog No. 501944759
Encompass_Preferred
Sparger assembly for 6l low profile Over Head Drive vessel. This fits through a 45mm sidearm. Designed for vertical side arm vessels. MADE IN THE USA Sparger tube: 10 1/8" long 316 l SST 20 ra material. 12mm od coarse glass frit (40-60µ) Gently introduces bubbles into vessel.
Catalog No. 50-194-4759 Supplier Bellco Glass, Inc. Supplier No. 196406415R
Sparger assembly for 6l low profile Over Head Drive vessel. This fits through a 45mm sidearm. Designed for vertical side arm vessels. MADE IN THE USA Sparger tube: 10 1/8" long 316 l SST 20 ra material. 12mm od coarse glass frit (40-60µ) Gently introduces bubbles into vessel.