Learn More
Bellco Glass, Inc. Low Profile 2-Port Assembly , 3L For Vertical Low Profile Vessels
SDP
Catalog No. 501944741
Encompass_Preferred
Low profile 45mm 2-port assembly for vertical low profile 3L vessels. MADE IN THE USA Seamed welded – electropolish 316l stainless steel material 32ra finish or better All tubes; .250"od x .152"id Siphon tube (LONG TUBE); 8 1/2" LONG. VENT TUBE: 1". SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600)
Catalog No. 50-194-4741 Supplier Bellco Glass, Inc. Supplier No. 196403402R
Low profile 45mm 2-port assembly for vertical low profile 3L vessels. MADE IN THE USA Seamed welded – electropolish 316l stainless steel material 32ra finish or better All tubes; .250"od x .152"id Siphon tube (LONG TUBE); 8 1/2" LONG. VENT TUBE: 1". SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600)