Learn More
Bellco Glass, Inc. Low Profile 2-Port Sparger, 3L For Low Profile Over Head Drive Vessels
SDP
Catalog No. 501944746
Encompass_Preferred
Sparger for 3l low profile Over Head Drive vessels. MADE IN THE USA To fit thru a 45mm sidearm Sparger tube: 8 1/2" long with glass frit.porosity: 12 c ( coarse) 12mm od coarse glass frit (40-60µ) Gently introduces bubbles into vessel 316l SST material 32ra (averasge)
Catalog No. 50-194-4746 Supplier Bellco Glass, Inc. Supplier No. 196403415R
Sparger for 3l low profile Over Head Drive vessels. MADE IN THE USA To fit thru a 45mm sidearm Sparger tube: 8 1/2" long with glass frit.porosity: 12 c ( coarse) 12mm od coarse glass frit (40-60µ) Gently introduces bubbles into vessel 316l SST material 32ra (averasge)