Learn More
Bellco Glass, Inc. Low Profile 2-Port Stainless Steel Assembly ,6L
SDP
Catalog No. 501944754
Encompass_Preferred
SST Low Profile Impeller Assembly For 6000mL Low Profile Vessels. Requires Over Head Drive & cap assembly. MADE IN THE USAMaterial: SST 316lConsist of: dust shield, shaft, paddle, locking ring, washer, cotter pinFour (4) SST set screws: 8-32 x 1/4"Dimensions: 9 1/2" to 12 1/2" oalPaddle size: 3" tall x 5 1/4" wideFully assembled
Catalog No. 50-194-4754 Supplier Bellco Glass, Inc. Supplier No. 196406402R
SST Low Profile Impeller Assembly For 6000mL Low Profile Vessels. Requires Over Head Drive & cap assembly. MADE IN THE USAMaterial: SST 316lConsist of: dust shield, shaft, paddle, locking ring, washer, cotter pinFour (4) SST set screws: 8-32 x 1/4"Dimensions: 9 1/2" to 12 1/2" oalPaddle size: 3" tall x 5 1/4" wideFully assembled