Learn More
Bellco Glass, Inc. Low Profile 3-Port Assembly , 3L For Vertical Low Profile Vessels
SDP
Catalog No. 501944742
Encompass_Preferred
Low profile 45mm 3-port assembly for vertical low profile 3L vessels. MADE IN THE USA Seam welded - electropolish 316l stainless steel material All tubes .250"od x .152"id Siphon tube: (long tube) 9 1/2", FILL TUBE: 3 13/16", VENT TUBE:2 5/32". SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600)
Catalog No. 50-194-4742 Supplier Bellco Glass, Inc. Supplier No. 196403403R
Low profile 45mm 3-port assembly for vertical low profile 3L vessels. MADE IN THE USA Seam welded - electropolish 316l stainless steel material All tubes .250"od x .152"id Siphon tube: (long tube) 9 1/2", FILL TUBE: 3 13/16", VENT TUBE:2 5/32". SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600)