Learn More
Bellco Glass, Inc. Low Profile 3-Port Harvest Assembly ,6L For Vertical Side Arm Vessels
SDP
Catalog No. 501944850
Encompass_Preferred
MADE IN THE USA. Material: 316l SST with electropolish finish, ra 32 or better, inside & outDimensions: harvest tube (3/8" od) - 1" above slug, 10 9/32" insertion lengthAddition tube (1/4"od) - 1" above slug, 2 21/64" belowVent tube (1/4" od) - 1" above slug, .975" below Supply with 45mm PP blue open top screw cap & white silicone gasket
Catalog No. 50-194-4850 Supplier Bellco Glass, Inc. Supplier No. 196463006
MADE IN THE USA. Material: 316l SST with electropolish finish, ra 32 or better, inside & outDimensions: harvest tube (3/8" od) - 1" above slug, 10 9/32" insertion lengthAddition tube (1/4"od) - 1" above slug, 2 21/64" belowVent tube (1/4" od) - 1" above slug, .975" below Supply with 45mm PP blue open top screw cap & white silicone gasket