Learn More
Bellco Glass, Inc. Low Profile 3-Port Sparger W/Holes,6L For Vertical Side Arm Vessels
SDP
Catalog No. 501944845
Encompass_Preferred
316-l stainless steel ra32 or better finish.040" drilled holes sparger for low profile vertical side arm vessels. MADE IN THE USA. Threaded end is removable for cleaning.Comes with 45mm blue polypropylene open top cap and silicone gasket.Triple tube configuration With Vent, Fill/Sample And Sparge.
Catalog No. 50-194-4845 Supplier Bellco Glass, Inc. Supplier No. 196462006
316-l stainless steel ra32 or better finish.040" drilled holes sparger for low profile vertical side arm vessels. MADE IN THE USA. Threaded end is removable for cleaning.Comes with 45mm blue polypropylene open top cap and silicone gasket.Triple tube configuration With Vent, Fill/Sample And Sparge.