Learn More
Bellco Glass, Inc. Low Profile 3-Port Stainless Steel Assembly , 6L
SDP
Catalog No. 501944755
Encompass_Preferred
Stainless steel 3-port assembly for media bottles, 6l low profile. Welded, 316l stainless steel construction, 1/4" inch od tubing. MADE IN THE USAElectropolished finish, continuous seal weldThree port model features siphon, fill and vent tubes45mm polypropylene screw cap and silicone gasket includedCap slug: 45mmTube specifications: 1/4" OD X 5/32" ID, SIPHON (LONG TUBE): 10 13/32" LONG BELOW CAP SLUG, FILL TUBE: 3" LONG BELOW CAP SLUG, VENT TUBE: 1" LONG BELOW CAP SLUG.
Catalog No. 50-194-4755 Supplier Bellco Glass, Inc. Supplier No. 196406403R
Stainless steel 3-port assembly for media bottles, 6l low profile. Welded, 316l stainless steel construction, 1/4" inch od tubing. MADE IN THE USAElectropolished finish, continuous seal weldThree port model features siphon, fill and vent tubes45mm polypropylene screw cap and silicone gasket includedCap slug: 45mmTube specifications: 1/4" OD X 5/32" ID, SIPHON (LONG TUBE): 10 13/32" LONG BELOW CAP SLUG, FILL TUBE: 3" LONG BELOW CAP SLUG, VENT TUBE: 1" LONG BELOW CAP SLUG.