Learn More
Medchemexpress LLC L-p-boronophenylalanine | 76410-58-7 | MFCD01075172 | 99.8% | 209.01 | C9H12BNO4 | 250 MG
SDP
Catalog No. 5000209229
Encompass_Preferred
L-p-Boronophenylalanine is a boron-containing amino acid used as a substrate for L-type amino acid transporters (LAT1 and LAT2). It selectively accumulates in tumor cells and is used in boron neutron capture therapy (BNCT) research and studies of amino acid transport in cancer biology.
  • High purity suitable for research applications.
  • Available as a white to off-white solid powder for storage stability.
  • Molecular formula C9H12BNO4 and molecular weight 209.01 g/mol.
  • Stable under recommended storage conditions (powder: -20°C for long-term storage).
Catalog No. 50-002-09229 Supplier Medchemexpress LLC Supplier No. 250MGHYW087830
L-p-Boronophenylalanine is a boron-containing amino acid used as a substrate for L-type amino acid transporters (LAT1 and LAT2). It selectively accumulates in tumor cells and is used in boron neutron capture therapy (BNCT) research and studies of amino acid transport in cancer biology.
  • High purity suitable for research applications.
  • Available as a white to off-white solid powder for storage stability.
  • Molecular formula C9H12BNO4 and molecular weight 209.01 g/mol.
  • Stable under recommended storage conditions (powder: -20°C for long-term storage).