Learn More
Bellco Glass, Inc. Low Profile Glass Impeller Assembly , 3L
SDP
Catalog No. 501944745
Encompass_Preferred
Low Profile Glass Impeller Assembly . MADE IN THE USA Size: 3000ml (for low profile 3l vessel)Paddle: 3" tall x 7" wide
Catalog No. 50-194-4745 Supplier Bellco Glass, Inc. Supplier No. 196403410
Low Profile Glass Impeller Assembly . MADE IN THE USA Size: 3000ml (for low profile 3l vessel)Paddle: 3" tall x 7" wide