Learn More
Bellco Glass, Inc. Low Profile Glass Impeller Assembly , 6L
SDP
Catalog No. 501944758
Encompass_Preferred
Low Profile Impeller Assembly. Size: for 6l low profile over head drive vessel. MADE IN THE USADimensions: 14" oal with compression fitting & capPaddle size: 3" tall x 7" wide
Catalog No. 50-194-4758 Supplier Bellco Glass, Inc. Supplier No. 196406410
Low Profile Impeller Assembly. Size: for 6l low profile over head drive vessel. MADE IN THE USADimensions: 14" oal with compression fitting & capPaddle size: 3" tall x 7" wide