Learn More
Bellco Glass, Inc. Low Profile Impeller Assembly ,3 L PTFE Paddle
SDP
Catalog No. 501944743
Encompass_Preferred
Ptfe impeller for use with over head drive motor and drive adapter in 3l low profile bioreactor vessel. MADE IN THE USA Paddle size: 3 x 6 3/4"
Catalog No. 50-194-4743 Supplier Bellco Glass, Inc. Supplier No. 196403404
Ptfe impeller for use with over head drive motor and drive adapter in 3l low profile bioreactor vessel. MADE IN THE USA Paddle size: 3 x 6 3/4"