Learn More
Bellco Glass, Inc. Low Profile PTFE Impeller Assembly ,6 L
SDP
Catalog No. 501944756
Encompass_Preferred
The Bellco low profile ptfe impellers are designed to be used with bellco low profile cell culture vessels. These low profile impeller consist of a semi-rigid ptfe blade, ptfe paddle holder and stainless steel shaft. Impellers are shipped fully assembled. Paddle Size: 3"Tx 6 3/4"W. Requires Bellco Overhead Drive & Cap Assembly. MADE IN THE USA
Catalog No. 50-194-4756 Supplier Bellco Glass, Inc. Supplier No. 196406404
The Bellco low profile ptfe impellers are designed to be used with bellco low profile cell culture vessels. These low profile impeller consist of a semi-rigid ptfe blade, ptfe paddle holder and stainless steel shaft. Impellers are shipped fully assembled. Paddle Size: 3"Tx 6 3/4"W. Requires Bellco Overhead Drive & Cap Assembly. MADE IN THE USA