Learn More
Bellco Glass, Inc. Low Profile Hot Shaker,230V
SDP
Catalog No. 501946179
Encompass_Preferred
Bellco’s new Low Profile Hot Shaker provides gentle, large orbit shaking motion combined with precise temperature control. Designed to be the perfect solution for incubation of electrophoretic gels and filter hybridization media, the Low Profile layout promotes easy access to the independent microprocessor driven temperature controller and reliable control of shaking speeds up to 75 oscillations per minute. The quiet brushless drive motor and repeatable, stable temperature settings from 5°C above ambient up to 99.9°C may be effectively used in most biochemical, chemical and microbiological applications.. MADE IN THE USA
Catalog No. 50-194-6179 Supplier Bellco Glass, Inc. Supplier No. 774642230
Add to Cart
Add to Cart
Request a Quote
Bellco’s new Low Profile Hot Shaker provides gentle, large orbit shaking motion combined with precise temperature control. Designed to be the perfect solution for incubation of electrophoretic gels and filter hybridization media, the Low Profile layout promotes easy access to the independent microprocessor driven temperature controller and reliable control of shaking speeds up to 75 oscillations per minute. The quiet brushless drive motor and repeatable, stable temperature settings from 5°C above ambient up to 99.9°C may be effectively used in most biochemical, chemical and microbiological applications.. MADE IN THE USA