Learn More
Bellco Glass, Inc. Low Profile Stainless Steel Impeller,3L
SDP
Catalog No. 501944740
Encompass_Preferred
Stainless steel 3L low profile impeller assembly For low profile Over Head Drive vessel. MADE IN THE USA Material: SST 316L Electropolish finish, ra 32 or better Fully assembled Paddle dimensions: 2" T X 5 1/4" W
Catalog No. 50-194-4740 Supplier Bellco Glass, Inc. Supplier No. 196403401
Stainless steel 3L low profile impeller assembly For low profile Over Head Drive vessel. MADE IN THE USA Material: SST 316L Electropolish finish, ra 32 or better Fully assembled Paddle dimensions: 2" T X 5 1/4" W