Learn More
Bellco Glass, Inc. Low Profile Thermowell Assembly , 3L For Low Profile Vessels
SDP
Catalog No. 501944744
Encompass_Preferred
45mm low profile Thermowell assembly for low profile 3l vessel. MADE IN THE USASeam welded - electropolish finish316l stainless steel material ra 32 finish or betterSupply with s/cap (a523-599) & white silicone liner (a523-600)1 - THERMOWELL TUBE: 5/16"OD X .242"ID X 1.0" above x 8 11/32" insertion 1- FILL TUBE: 1/4"OD X .152"ID X 1.0" above x 8 11/32" insertion 1- VENT TUBE: .250"OD X .152"ID X 1.0"LONG
Catalog No. 50-194-4744 Supplier Bellco Glass, Inc. Supplier No. 196403405
45mm low profile Thermowell assembly for low profile 3l vessel. MADE IN THE USASeam welded - electropolish finish316l stainless steel material ra 32 finish or betterSupply with s/cap (a523-599) & white silicone liner (a523-600)1 - THERMOWELL TUBE: 5/16"OD X .242"ID X 1.0" above x 8 11/32" insertion 1- FILL TUBE: 1/4"OD X .152"ID X 1.0" above x 8 11/32" insertion 1- VENT TUBE: .250"OD X .152"ID X 1.0"LONG