Learn More
Bellco Glass, Inc. Low Profile Thermowell Assembly , 6L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944757
Encompass_Preferred
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. MADE IN THE USA 45mm THERMOWELL ASSY FOR L/P 6L VESSEL, SEAM WELDED - ELECTROPOLISH FINISH, 316L SST MATERIAL, SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600), 1- THERMOWELL TUBE: 5/16"OD X .242"ID X 1.0" above x10 9/32" insertion, 1- SIPHON TUBE: 1/4"OD X 5/32" ID X 1.0" above x 10 9/32" insertion, 1- VENT TUBE: 1/4"OD X 5/32" ID X 1.0".
Catalog No. 50-194-4757 Supplier Bellco Glass, Inc. Supplier No. 196406405
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. MADE IN THE USA 45mm THERMOWELL ASSY FOR L/P 6L VESSEL, SEAM WELDED - ELECTROPOLISH FINISH, 316L SST MATERIAL, SUPPLY WITH S/CAP (A523-599) & WHITE SILICONE LINER (A523-600), 1- THERMOWELL TUBE: 5/16"OD X .242"ID X 1.0" above x10 9/32" insertion, 1- SIPHON TUBE: 1/4"OD X 5/32" ID X 1.0" above x 10 9/32" insertion, 1- VENT TUBE: 1/4"OD X 5/32" ID X 1.0".