Learn More
Bellco Glass, Inc. Low Profile Vessel W/ 4 Side Arms, 3L
SDP
Supplier:  Bellco Glass, Inc. 196403400
Encompass_Preferred
LOW PROFILE CELL CULTURE VESSELS. Reduced height vessels maximize incubator space. This vessel is suitable for all cell culture applications (insect, plant, & bacterial). 100mm neck takes drive adapter and overhead drive motor. 45MM vertical sidearm accept port and thermowell assemblies or probes. Supplied with 100mm polypropylene center cap, sidearm cap and pouring rings. OD X OAH (MM): 240 X 250. USE 8L FLASK CARRIER & 1964-51040 HEATING BLANKET . MADE IN THE USA
Catalog No. 50-194-4739