Learn More
Bellco Glass, Inc. Low Profile Vessel W/4 Side Arms, 6L
SDP
Catalog No. 501944752
Encompass_Preferred
LOW PROFILE CELL CULTURE VESSELS. REDUCED HEIGHT VESSELS MAXIMIZE INCUBATOR SPACE SUITABLE FOR ALL CELL CULTURE APPLICATIONS-INSECT, PLANT,BACTERIAL 100MM NECK FOR CAP ASSEMBLY AND OVERHEAD DRIVE 45MM VERTICAL SIDEARM ACCEPT PORT AND THERMOWELL ASSEMBLIES OR PROBES. MADE IN THE USA -SUPPLIED WITH 100MM CENTER CAP, 45MM CAPS AND POURING RINGS OD X OAH (MM): 240 X 295 Qty: 1 ea.
Catalog No. 50-194-4752 Supplier Bellco Glass, Inc. Supplier No. 196406400
LOW PROFILE CELL CULTURE VESSELS. REDUCED HEIGHT VESSELS MAXIMIZE INCUBATOR SPACE SUITABLE FOR ALL CELL CULTURE APPLICATIONS-INSECT, PLANT,BACTERIAL 100MM NECK FOR CAP ASSEMBLY AND OVERHEAD DRIVE 45MM VERTICAL SIDEARM ACCEPT PORT AND THERMOWELL ASSEMBLIES OR PROBES. MADE IN THE USA -SUPPLIED WITH 100MM CENTER CAP, 45MM CAPS AND POURING RINGS OD X OAH (MM): 240 X 295 Qty: 1 ea.