Learn More
Bellco Glass, Inc. Low Profile Vessel w/6 Side Arms, 3L
SDP
Catalog No. 501944749
Encompass_Preferred
This vessel is suitable for all cell culture applications- insect, plant and bacterial. Reduced height vessels maximize incubator space. Low profile cell culture vessels. MADE IN THE USAReduced height vessels maximize incubator space.Suitable for all cell culture applications-insect, plant, and bacterial.100mm neck for cap assembly and overhead drive.45mm vertical sidearm accept port and thermowell assemblies or probes.Supplied with 100mm center cap, 45mm caps and pouring rings.Od x oah (mm): 240 x 250.Use 8l flask carrier & 1964-51040 heating blanket.
Catalog No. 50-194-4749 Supplier Bellco Glass, Inc. Supplier No. 196403600
This vessel is suitable for all cell culture applications- insect, plant and bacterial. Reduced height vessels maximize incubator space. Low profile cell culture vessels. MADE IN THE USAReduced height vessels maximize incubator space.Suitable for all cell culture applications-insect, plant, and bacterial.100mm neck for cap assembly and overhead drive.45mm vertical sidearm accept port and thermowell assemblies or probes.Supplied with 100mm center cap, 45mm caps and pouring rings.Od x oah (mm): 240 x 250.Use 8l flask carrier & 1964-51040 heating blanket.