Learn More
Bellco Glass, Inc. Low Profile Vessel W/6 Side Arms,6L
SDP
Catalog No. 501944762
Encompass_Preferred
Reduced height vessels maximize incubator space. This vessel is suitable for all cell culture applications (Insect, Plant, & Bacterial). 100mm neck takes drive adapter and overhead drive motor. Supplied with 100mm polypropylene center cap, sidearm cap and pouring ring. MADE IN THE USASpecifications:45mm Vertical Sidearm Accepts Port And Thermowell Assemblies Or ProbesOD x OAL: (mm): 240×295Use 8L Flask Carrier & 1964-51040 Heating Blanket
Catalog No. 50-194-4762 Supplier Bellco Glass, Inc. Supplier No. 196406600
Reduced height vessels maximize incubator space. This vessel is suitable for all cell culture applications (Insect, Plant, & Bacterial). 100mm neck takes drive adapter and overhead drive motor. Supplied with 100mm polypropylene center cap, sidearm cap and pouring ring. MADE IN THE USASpecifications:45mm Vertical Sidearm Accepts Port And Thermowell Assemblies Or ProbesOD x OAL: (mm): 240×295Use 8L Flask Carrier & 1964-51040 Heating Blanket