Learn More
Medchemexpress LLC HY-113147A 5mg Medchemexpress, L-Palmitoylcarnitine (chloride) CAS:18877-64-0 Purity:>98%
SDP
Catalog No. 502080840
Encompass_Preferred
Medchemexpress, HY-113147A 5mg L-Palmitoylcarnitine (chloride) CAS:18877-64-0 L-Palmitoylcarnitine chloride, a long-chain acylcarnitine and a fatty acid metabolite, accumulates in the sarcolemma and deranges the membrane lipid environment during ischaemia. L-Palmitoylcarnitine chloride inhibits KATP channel activity, without affecting the single channel conductance, through interaction with Kir6.2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0840 Supplier Medchemexpress LLC Supplier No. HY113147A5MG
Medchemexpress, HY-113147A 5mg L-Palmitoylcarnitine (chloride) CAS:18877-64-0 L-Palmitoylcarnitine chloride, a long-chain acylcarnitine and a fatty acid metabolite, accumulates in the sarcolemma and deranges the membrane lipid environment during ischaemia. L-Palmitoylcarnitine chloride inhibits KATP channel activity, without affecting the single channel conductance, through interaction with Kir6.2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.