Learn More
Medchemexpress LLC HY-113147B 10mg Medchemexpress, L-Palmitoylcarnitine (TFA) CAS: Purity:>98%
SDP
Catalog No. 502080841
Encompass_Preferred
Medchemexpress, HY-113147B 10mg L-Palmitoylcarnitine (TFA) CAS: L-Palmitoylcarnitine TFA, a long-chain acylcarnitine and a fatty acid metabolite, accumulates in the sarcolemma and deranges the membrane lipid environment during ischaemia. L-Palmitoylcarnitine TFA inhibits KATP channel activity, without affecting the single channel conductance, through interaction with Kir6.2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0841 Supplier Medchemexpress LLC Supplier No. HY113147B10MG
Medchemexpress, HY-113147B 10mg L-Palmitoylcarnitine (TFA) CAS: L-Palmitoylcarnitine TFA, a long-chain acylcarnitine and a fatty acid metabolite, accumulates in the sarcolemma and deranges the membrane lipid environment during ischaemia. L-Palmitoylcarnitine TFA inhibits KATP channel activity, without affecting the single channel conductance, through interaction with Kir6.2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.