Learn More
Medchemexpress LLC L-Palmitoylcarnitine TFA | 98.0% | 513.63 | 50 MG
SDP
Catalog No. 5000436021
Encompass_Preferred
L-Palmitoylcarnitine TFA is a long-chain acylcarnitine and a fatty acid metabolite that accumulates in the sarcolemma and deranges the membrane lipid environment during ischemia. It inhibits KATP channel activity, without affecting single channel conductance, through interaction with Kir6.2. L-Palmitoylcarnitine (1 μM) enhances the ATP sensitivity of the channel (IC50 fell from 62 to 30 μM).
Catalog No. 50-004-36021 Supplier Medchemexpress LLC Supplier No. HY113147B50MG
L-Palmitoylcarnitine TFA is a long-chain acylcarnitine and a fatty acid metabolite that accumulates in the sarcolemma and deranges the membrane lipid environment during ischemia. It inhibits KATP channel activity, without affecting single channel conductance, through interaction with Kir6.2. L-Palmitoylcarnitine (1 μM) enhances the ATP sensitivity of the channel (IC50 fell from 62 to 30 μM).