Learn More
Medchemexpress LLC L-Perillaldehyde | 18031-40-8 | 150.22 | 10 G
SDP
Catalog No. 5000415266
Encompass_Preferred
L-Perillaldehyde is a major component found in the essential oil of Perillae Herba. Inhalation of L-Perillaldehyde has shown antidepressant-like activity through its effect on the olfactory nervous function. It is intended for research use only and is not sold to patients.
  • Major component in the essential oil of Perillae Herba.
  • Shows antidepressant-like activity through olfactory nervous function when inhaled.
  • Reduces immobility in stress-induced depression-like model mice during forced swimming tests at dosages of 0.0965 and 0.965 mg/mouse/day via inhalation.
Catalog No. 50-004-15266 Supplier Medchemexpress LLC Supplier No. HYN208510G
L-Perillaldehyde is a major component found in the essential oil of Perillae Herba. Inhalation of L-Perillaldehyde has shown antidepressant-like activity through its effect on the olfactory nervous function. It is intended for research use only and is not sold to patients.
  • Major component in the essential oil of Perillae Herba.
  • Shows antidepressant-like activity through olfactory nervous function when inhaled.
  • Reduces immobility in stress-induced depression-like model mice during forced swimming tests at dosages of 0.0965 and 0.965 mg/mouse/day via inhalation.