Learn More
Medchemexpress LLC L-Perillaldehyde | 18031-40-8 | 150.22 | 250G
SDP
Catalog No. 5000389521
Encompass_Preferred
L-Perillaldehyde is a primary constituent found in the essential oil of Perillae Herba. Studies indicate that the inhalation of L-Perillaldehyde exhibits antidepressant-like properties through its interaction with the olfactory nervous system.
  • Animal model: Stress-induced depression-like model mice
  • Dosage: 0.0965 and 0.965 mg/mouse/day
  • Administration: Inhalation
  • Result: Reduced immobility in the forced swimming test
Catalog No. 50-003-89521 Supplier Medchemexpress LLC Supplier No. 250GHYN2085
L-Perillaldehyde is a primary constituent found in the essential oil of Perillae Herba. Studies indicate that the inhalation of L-Perillaldehyde exhibits antidepressant-like properties through its interaction with the olfactory nervous system.
  • Animal model: Stress-induced depression-like model mice
  • Dosage: 0.0965 and 0.965 mg/mouse/day
  • Administration: Inhalation
  • Result: Reduced immobility in the forced swimming test