Learn More
Medchemexpress LLC L-Perillaldehyde | 18031-40-8 | 150.22 | 50 G
SDP
Catalog No. 5000415267
Encompass_Preferred
L-Perillaldehyde is a major component found in the essential oil of Perillae Herba. It has demonstrated antidepressant-like activity through the olfactory nervous function when inhaled. This product is intended for research use only and is not sold to patients.
  • High purity of at least 97.0%.
  • Appears as a colorless to light yellow liquid.
  • Classified as a terpenoid.
  • Derived from plants such as Leguminosae and Labiatae.
  • Shipped at room temperature within the continental US.
  • Storage at -20°C under nitrogen. In solvent, store at -80°C for 6 months or -20°C for 1 month (under nitrogen).
  • Inhalation exhibits antidepressant-like activity.
Catalog No. 50-004-15267 Supplier Medchemexpress LLC Supplier No. HYN208550G
L-Perillaldehyde is a major component found in the essential oil of Perillae Herba. It has demonstrated antidepressant-like activity through the olfactory nervous function when inhaled. This product is intended for research use only and is not sold to patients.
  • High purity of at least 97.0%.
  • Appears as a colorless to light yellow liquid.
  • Classified as a terpenoid.
  • Derived from plants such as Leguminosae and Labiatae.
  • Shipped at room temperature within the continental US.
  • Storage at -20°C under nitrogen. In solvent, store at -80°C for 6 months or -20°C for 1 month (under nitrogen).
  • Inhalation exhibits antidepressant-like activity.