Learn More
Medchemexpress LLC L-Perillaldehyde | 18031-40-8 | 150.22 | 500 G
SDP
Catalog No. 5000414972
Encompass_Preferred
L-Perillaldehyde is a major component in the essential oil found in Perillae Herba. Inhalation of L-Perillaldehyde demonstrates antidepressant-like activity through the olfactory nervous function.
  • Major component in essential oil found in Perillae Herba
  • Demonstrates antidepressant-like activity
  • Acts through olfactory nervous function
  • Reduces immobility in stress-induced depression-like model mice
Catalog No. 50-004-14972 Supplier Medchemexpress LLC Supplier No. HYN2085500G
L-Perillaldehyde is a major component in the essential oil found in Perillae Herba. Inhalation of L-Perillaldehyde demonstrates antidepressant-like activity through the olfactory nervous function.
  • Major component in essential oil found in Perillae Herba
  • Demonstrates antidepressant-like activity
  • Acts through olfactory nervous function
  • Reduces immobility in stress-induced depression-like model mice