Learn More
Medchemexpress LLC L-Perillaldehyde | 18031-40-8 | MFCD00001543 | 150.22 | 5 G
SDP
Catalog No. 5000415734
Encompass_Preferred
L-Perillaldehyde is a primary component found in the essential oil within Perillae Herba. Inhalation of L-Perillaldehyde has been observed to exhibit antidepressant-like effects through its interaction with the olfactory nervous system.
  • Major component in the essential oil of Perillae Herba.
  • Shows antidepressant-like activity through olfactory nervous function upon inhalation.
  • Purity: ≥97.0%.
Catalog No. 50-004-15734 Supplier Medchemexpress LLC Supplier No. HYN20855G
L-Perillaldehyde is a primary component found in the essential oil within Perillae Herba. Inhalation of L-Perillaldehyde has been observed to exhibit antidepressant-like effects through its interaction with the olfactory nervous system.
  • Major component in the essential oil of Perillae Herba.
  • Shows antidepressant-like activity through olfactory nervous function upon inhalation.
  • Purity: ≥97.0%.