Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Phenylalanine-α,β,β,2,3,4,5,6-d8 | 17942-32-4 | 99.6% | 173.24 | 5 MG
SDP

Catalog No. 5000428084
Encompass_Preferred

L-Phenylalanine-d8 is the deuterium labeled L-Phenylalanine, an essential amino acid isolated from Escherichia coli. It acts as an α2δ subunit of voltage-dependent Ca+ channels antagonist with a Ki of 980 nM. L-phenylalanine is also a competitive antagonist for the glycine- and glutamate-binding sites of N-methyl-D-aspartate receptors (NMDARs) (KB of 573 μM) and non-NMDARs, respectively. It is widely used in the production of food flavors and pharmaceuticals.

  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Appearance: solid, white to off-white.
  • Powder storage: -20°C for 3 years or 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-28084 Supplier Medchemexpress LLC Supplier No. HYN0215S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Phenylalanine-d8 is the deuterium labeled L-Phenylalanine, an essential amino acid isolated from Escherichia coli. It acts as an α2δ subunit of voltage-dependent Ca+ channels antagonist with a Ki of 980 nM. L-phenylalanine is also a competitive antagonist for the glycine- and glutamate-binding sites of N-methyl-D-aspartate receptors (NMDARs) (KB of 573 μM) and non-NMDARs, respectively. It is widely used in the production of food flavors and pharmaceuticals.

  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Appearance: solid, white to off-white.
  • Powder storage: -20°C for 3 years or 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.