Learn More
Medchemexpress LLC L-phenylalanine, 3-[[2,4-dicyano-3-[4-[2-(methylamino)-2-oxoethoxy]phenyl]pyrido[1,2-a]benzimi | 1941174-13-5 | 99.8% | 50 MG
SDP
Catalog No. 5000232712
Encompass_Preferred
KMH-233 is a small-molecule LAT1 (SLC7A5) inhibitor used in biochemical and cellular research to investigate amino-acid transport and cancer metabolism.
  • Potent, reversible, selective LAT1 inhibitor.
  • High purity suitable for research (>99.7%).
  • Molecular weight 587.58 g·mol⁻1 and formula C32H25N7O5.
  • Solid form, typically supplied in 50 MG quantities.
  • Applicable to transporter assays, cellular studies, and cancer metabolism research.
Catalog No. 50-002-32712 Supplier Medchemexpress LLC Supplier No. HY12013950MG
KMH-233 is a small-molecule LAT1 (SLC7A5) inhibitor used in biochemical and cellular research to investigate amino-acid transport and cancer metabolism.
  • Potent, reversible, selective LAT1 inhibitor.
  • High purity suitable for research (>99.7%).
  • Molecular weight 587.58 g·mol⁻1 and formula C32H25N7O5.
  • Solid form, typically supplied in 50 MG quantities.
  • Applicable to transporter assays, cellular studies, and cancer metabolism research.